The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080927.3 | NP_563615.3 | 7 - 8 | 1227 | 75 |
XM_011512419.2 | XP_011510721.1 | 6 - 7 | 943 | 75 | |
GenBank mRNA | AB073146.1 | - | 7 - 8 | 1227 | 75 |
AF387547.1 | - | 7 - 8 | 875 | 75 | |
AK056350.1 | - | 7 - 8 | 1220 | 75 | |
AK302649.1 | - | 7 - 8 | 707 | 75 | |
BC029658.1 | - | 7 - 8 | 746 | 75 |