Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FREM1
Entrez Gene ID
158326
Gene Name
FRAS1 related extracellular matrix 1
Gene Aliases
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
Chromosome Location
Chr.9: 14734666 - 14911653 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177704.1 NP_001171175.1 12 - 13 1953 58
NM_144966.5 NP_659403.4 36 - 37 7072 58
XM_005251382.3 XP_005251439.1 35 - 36 7320 58
XM_005251384.4 XP_005251441.1 14 - 15 2366 58
XM_006716729.3 XP_006716792.1 15 - 16 2611 58
XM_011517758.2 XP_011516060.1 12 - 13 2002 58
XM_017014316.1 XP_016869805.1 35 - 36 7347 58
XM_017014317.1 XP_016869806.1 36 - 37 6890 58
XM_017014318.1 XP_016869807.1 36 - 37 7645 58
XM_017014319.1 XP_016869808.1 37 - 38 7549 58
XM_017014320.1 XP_016869809.1 36 - 37 7476 58
XM_017014321.1 XP_016869810.1 35 - 36 7139 58
XM_017014322.1 XP_016869811.1 35 - 36 6933 58
XM_017014323.1 XP_016869812.1 35 - 36 6606 58
XM_017014324.1 XP_016869813.1 34 - 35 7197 58
XM_017014326.1 XP_016869815.1 34 - 35 6163 58
XM_017014327.1 XP_016869816.1 31 - 32 5532 58
GenBank mRNA AB160987.1 - 34 - 35 6256 58
AK058190.1 - 9 - 10 1122 58
BC031064.1 - 12 - 13 1953 58
BU677942.1 - 2 - 3 58
BX537574.1 - 29 - 30 5647 58
BX641104.1 - 6 - 7 6577 58

View Full Product Details