Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAUS8
Entrez Gene ID
93323
Gene Name
HAUS augmin like complex subunit 8
Gene Aliases
DGT4, HICE1, NY-SAR-48
Chromosome Location
Chr.19: 17049761 - 17075533 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011699.1 NP_001011699.1 10 - 11 1038 152
NM_033417.1 NP_219485.1 10 - 11 1041 152
XM_005260154.4 XP_005260211.1 9 - 10 874 152
GenBank mRNA AK295994.1 - 10 - 11 1113 152
BC004398.1 - 10 - 11 928 152
BC010176.2 - 4 - 5 477 152

View Full Product Details