The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164232.1 | NP_001157704.1 | 6 - 7 | 917 | 76 |
NM_015214.2 | NP_056029.2 | 6 - 7 | 1094 | 76 | |
XM_005273454.2 | XP_005273511.1 | 6 - 7 | 1242 | 76 | |
XM_005273455.3 | XP_005273512.1 | 6 - 7 | 1242 | 76 | |
XM_011544456.1 | XP_011542758.1 | 6 - 7 | 1242 | 76 | |
XM_017013252.1 | XP_016868741.1 | 6 - 7 | 1051 | 76 | |
XM_017013253.1 | XP_016868742.1 | 6 - 7 | 1242 | 76 | |
XM_017013254.1 | XP_016868743.1 | 6 - 7 | 899 | 76 | |
XM_017013256.1 | XP_016868745.1 | 6 - 7 | 998 | 76 | |
GenBank mRNA | AB018268.1 | - | 3 - 4 | 304 | 76 |
AK023218.1 | - | 2 - 3 | 123 | 76 | |
AK056525.1 | - | 6 - 7 | 996 | 76 | |
AK125904.1 | - | 6 - 7 | 917 | 76 | |
AK299875.1 | - | 4 - 5 | 592 | 76 | |
BC010504.1 | - | 1 - 2 | 65 | 76 |