Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDCA5
Entrez Gene ID
113130
Gene Name
cell division cycle associated 5
Gene Aliases
SORORIN
Chromosome Location
Chr.11: 65061009 - 65084143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080668.3 NP_542399.1 5 - 6 841 72
GenBank mRNA AK093670.1 - 1 - 2 416 72
AK291490.1 - 5 - 6 765 72
AL832526.1 - 4 - 5 558 72
BC011000.2 - 5 - 6 749 72

View Full Product Details