The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144628.3 | NP_653229.1 | 5 - 6 | 776 | 63 |
NR_111901.1 | 5 - 6 | 776 | 63 | ||
XM_005260661.1 | XP_005260718.1 | 5 - 6 | 766 | 63 | |
XM_006723540.3 | XP_006723603.1 | 4 - 5 | 555 | 63 | |
XM_017027645.1 | XP_016883134.1 | 4 - 5 | 547 | 63 | |
GenBank mRNA | AB449906.1 | - | 5 - 6 | 628 | 63 |
AK095110.1 | - | 4 - 5 | 935 | 63 | |
AK127062.1 | - | 5 - 6 | 756 | 63 | |
AK291648.1 | - | 5 - 6 | 739 | 63 | |
BC014983.1 | - | 5 - 6 | 726 | 63 | |
DA230581.1 | - | 2 - 3 | 289 | 63 |