The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301010.1 | NP_001287939.1 | 5 - 6 | 702 | 108 |
NM_138775.2 | NP_620130.2 | 5 - 6 | 731 | 108 | |
XM_005271728.2 | XP_005271785.1 | 5 - 6 | 726 | 108 | |
XM_005271729.3 | XP_005271786.1 | 5 - 6 | 699 | 108 | |
XM_017018553.1 | XP_016874042.1 | 5 - 6 | 704 | 108 | |
XM_017018554.1 | XP_016874043.1 | 5 - 6 | 736 | 108 | |
XM_017018555.1 | XP_016874044.1 | 5 - 6 | 731 | 108 | |
XM_017018556.1 | XP_016874045.1 | 5 - 6 | 726 | 108 | |
XM_017018557.1 | XP_016874046.1 | 5 - 6 | 704 | 108 | |
GenBank mRNA | AB218768.1 | - | 5 - 6 | 731 | 108 |
AK293603.1 | - | 5 - 6 | 665 | 108 | |
AK298800.1 | - | 3 - 4 | 291 | 108 | |
AK304413.1 | - | 5 - 6 | 730 | 108 | |
BC015183.1 | - | 5 - 6 | 697 | 108 |