The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014991.4 | NP_055806.2 | 64 - 65 | 10229 | 81 |
XM_005262858.4 | XP_005262915.1 | 66 - 67 | 10270 | 81 | |
XM_011531757.2 | XP_011530059.1 | 65 - 66 | 10181 | 81 | |
XM_011531759.2 | XP_011530061.1 | 64 - 65 | 10143 | 81 | |
XM_011531760.2 | XP_011530062.1 | 64 - 65 | 10081 | 81 | |
XM_011531761.2 | XP_011530063.1 | 65 - 66 | 10136 | 81 | |
XM_011531762.2 | XP_011530064.1 | 64 - 65 | 10130 | 81 | |
XM_011531763.2 | XP_011530065.1 | 63 - 64 | 10027 | 81 | |
XM_011531764.2 | XP_011530066.1 | 64 - 65 | 10097 | 81 | |
XM_011531765.2 | XP_011530067.1 | 61 - 62 | 9832 | 81 | |
XM_017007906.1 | XP_016863395.1 | 67 - 68 | 10398 | 81 | |
XM_017007907.1 | XP_016863396.1 | 67 - 68 | 10395 | 81 | |
XM_017007908.1 | XP_016863397.1 | 66 - 67 | 10327 | 81 | |
XM_017007909.1 | XP_016863398.1 | 52 - 53 | 8023 | 81 | |
GenBank mRNA | AB023210.2 | - | 29 - 30 | 3912 | 81 |
AF538685.1 | - | 63 - 64 | 10121 | 81 | |
BC013377.2 | - | 1 - 2 | 573 | 81 | |
BC015214.1 | - | 4 - 5 | 555 | 81 | |
BC065502.1 | - | 11 - 12 | 1579 | 81 | |
BC119633.2 | - | 1 - 2 | 276 | 81 | |
BC148294.1 | - | 1 - 2 | 276 | 81 | |
BC148298.2 | - | 1 - 2 | 276 | 81 | |
BC150246.1 | - | 29 - 30 | 3933 | 81 | |
BX648077.1 | - | 21 - 22 | 2794 | 81 | |
BX648153.1 | - | 24 - 25 | 3122 | 81 |