Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCEAL7
Entrez Gene ID
56849
Gene Name
transcription elongation factor A like 7
Gene Aliases
MPMGp800C04260Q003, WEX5
Chromosome Location
Chr.X: 103330186 - 103332326 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152278.3 NP_689491.1 1 - 2 119 119
GenBank mRNA AJ297363.1 - 1 - 2 75 119
BC005988.1 - 1 - 2 65 119
BC016786.1 - 1 - 2 70 119
DB481725.1 - 1 - 2 119 119

View Full Product Details