The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320653.1 | NP_001307582.1 | 1 - 2 | 386 | 109 |
NM_002532.5 | NP_002523.2 | 1 - 2 | 386 | 109 | |
GenBank mRNA | AK225247.1 | - | 1 - 2 | 309 | 109 |
AK298158.1 | - | 2 - 3 | 163 | 109 | |
AK299000.1 | - | 1 - 2 | 309 | 109 | |
AK308142.1 | - | 1 - 2 | 198 | 109 | |
BC000335.2 | - | 1 - 2 | 299 | 109 | |
BI552947.1 | - | 1 - 2 | 328 | 109 | |
Y08612.2 | - | 1 - 2 | 345 | 109 |