The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002556.2 | NP_002547.1 | 11 - 12 | 2357 | 115 |
GenBank mRNA | AF185696.1 | - | 11 - 12 | 2357 | 115 |
AK299071.1 | - | 7 - 8 | 801 | 115 | |
AK310538.1 | - | 5 - 6 | 615 | 115 | |
BC011581.1 | - | 11 - 12 | 1945 | 115 | |
BC063121.1 | - | 5 - 6 | 716 | 115 | |
M86917.1 | - | 11 - 12 | 2357 | 115 |