The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006482.2 | NP_006473.2 | 2 - 3 | 599 | 74 |
GenBank mRNA | AK313937.1 | - | 2 - 3 | 611 | 74 |
BC006375.2 | - | 3 - 4 | 410 | 74 | |
Y13493.1 | - | 2 - 3 | 357 | 74 |