The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004947.4 | NP_004938.1 | 43 - 44 | 4525 | 79 |
XM_005264914.3 | XP_005264971.1 | 43 - 44 | 4797 | 79 | |
XM_005264915.3 | XP_005264972.1 | 43 - 44 | 4797 | 79 | |
XM_005264916.3 | XP_005264973.1 | 42 - 43 | 4548 | 79 | |
XM_005264917.3 | XP_005264974.1 | 43 - 44 | 4755 | 79 | |
XM_005264918.3 | XP_005264975.1 | 42 - 43 | 4427 | 79 | |
XM_006713008.3 | XP_006713071.1 | 43 - 44 | 4797 | 79 | |
XM_006713009.3 | XP_006713072.1 | 42 - 43 | 4710 | 79 | |
XM_006713010.3 | XP_006713073.1 | 43 - 44 | 4560 | 79 | |
XM_011533441.2 | XP_011531743.1 | 32 - 33 | 5406 | 79 | |
XM_011533443.2 | XP_011531745.1 | 19 - 20 | 3087 | 79 | |
XM_011533444.2 | XP_011531746.1 | 19 - 20 | 3011 | 79 | |
XM_011533445.2 | XP_011531747.1 | 19 - 20 | 3032 | 79 | |
XM_017005825.1 | XP_016861314.1 | 43 - 44 | 4576 | 79 | |
XM_017005826.1 | XP_016861315.1 | 43 - 44 | 4566 | 79 | |
XM_017005827.1 | XP_016861316.1 | 34 - 35 | 3717 | 79 | |
XM_017005828.1 | XP_016861317.1 | 18 - 19 | 2442 | 79 | |
GenBank mRNA | AB002297.1 | - | 38 - 39 | 4133 | 79 |
AY145303.1 | - | 43 - 44 | 4502 | 79 | |
AY254099.1 | - | 43 - 44 | 4525 | 79 | |
BC131746.1 | - | 8 - 9 | 893 | 79 |