Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRUNE2
Entrez Gene ID
158471
Gene Name
prune homolog 2
Gene Aliases
BMCC1, BNIPXL, C9orf65, KIAA0367
Chromosome Location
Chr.9: 76611376 - 76906220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308047.1 NP_001294976.1 15 - 16 9093 113
NM_001308048.1 NP_001294977.1 15 - 16 9096 113
NM_001308049.1 NP_001294978.1 7 - 8 1171 113
NM_001308050.1 NP_001294979.1 6 - 7 1026 113
NM_001308051.1 NP_001294980.1 6 - 7 1029 113
NM_015225.2 NP_056040.2 14 - 15 9084 113
NR_131751.1 13 - 14 8969 113
XM_005251745.1 XP_005251802.1 15 - 16 9229 113
XM_005251746.1 XP_005251803.1 15 - 16 9229 113
XM_005251748.1 XP_005251805.1 15 - 16 9226 113
XM_005251750.1 XP_005251807.1 14 - 15 9220 113
XM_005251751.1 XP_005251808.1 15 - 16 9229 113
XM_005251754.1 XP_005251811.1 14 - 15 9220 113
XM_006716982.1 XP_006717045.1 15 - 16 9229 113
XM_006716983.1 XP_006717046.1 15 - 16 9226 113
XM_006716984.2 XP_006717047.1 15 - 16 9229 113
XM_006716985.1 XP_006717048.1 14 - 15 9220 113
XM_006716986.1 XP_006717049.1 14 - 15 9217 113
XM_011518323.1 XP_011516625.1 14 - 15 9220 113
XM_011518326.2 XP_011516628.1 15 - 16 9229 113
XM_011518327.1 XP_011516629.1 15 - 16 9229 113
XM_011518328.2 XP_011516630.1 15 - 16 9229 113
XM_017014345.1 XP_016869834.1 15 - 16 9226 113
XM_017014346.1 XP_016869835.1 14 - 15 9217 113
XM_017014347.1 XP_016869836.1 15 - 16 9226 113
XM_017014348.1 XP_016869837.1 14 - 15 9220 113
XM_017014349.1 XP_016869838.1 14 - 15 9217 113
XM_017014350.1 XP_016869839.1 15 - 16 9226 113
XM_017014351.1 XP_016869840.1 14 - 15 9220 113
XM_017014352.1 XP_016869841.1 14 - 15 9217 113
XM_017014353.1 XP_016869842.1 15 - 16 9229 113
XM_017014354.1 XP_016869843.1 6 - 7 818 113
XM_017014355.1 XP_016869844.1 6 - 7 818 113
XM_017014356.1 XP_016869845.1 6 - 7 818 113
XM_017014357.1 XP_016869846.1 6 - 7 818 113
XM_017014358.1 XP_016869847.1 6 - 7 2672 113
XM_017014359.1 XP_016869848.1 6 - 7 818 113
GenBank mRNA AB002365.2 - 7 - 8 2156 113
AB050197.1 - 8 - 9 8190 113
AB535152.1 - 15 - 16 8969 113
AK296269.1 - 7 - 8 953 113
AK298805.1 - 6 - 7 808 113
AK299857.1 - 6 - 7 811 113
AL834213.1 - 8 - 9 7260 113
AY439213.1 - 8 - 9 2165 113
AY439214.1 - 8 - 9 2165 113
BC150641.1 - 8 - 9 8102 113
CB146736.1 - 4 - 5 342 113
FJ808772.1 - 15 - 16 9032 113
FJ808773.1 - 13 - 14 8908 113

View Full Product Details