The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015284.3 | NP_056099.3 | 54 - 55 | 7659 | 62 |
XM_005270686.3 | XP_005270743.1 | 56 - 57 | 7891 | 62 | |
XM_006710501.3 | XP_006710564.1 | 55 - 56 | 7778 | 62 | |
XM_011541106.2 | XP_011539408.1 | 55 - 56 | 7929 | 62 | |
XM_011541107.2 | XP_011539409.1 | 52 - 53 | 7426 | 62 | |
XM_017000819.1 | XP_016856308.1 | 56 - 57 | 7738 | 62 | |
XM_017000820.1 | XP_016856309.1 | 55 - 56 | 7720 | 62 | |
XM_017000821.1 | XP_016856310.1 | 52 - 53 | 7412 | 62 | |
GenBank mRNA | AB007936.2 | - | 40 - 41 | 5511 | 62 |
BC151232.1 | - | 40 - 41 | 5511 | 62 |