The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015284.3 | NP_056099.3 | 69 - 70 | 9998 | 73 |
XM_005270686.3 | XP_005270743.1 | 71 - 72 | 10230 | 73 | |
XM_006710501.3 | XP_006710564.1 | 70 - 71 | 10117 | 73 | |
XM_011541106.2 | XP_011539408.1 | 70 - 71 | 10268 | 73 | |
XM_011541107.2 | XP_011539409.1 | 67 - 68 | 9765 | 73 | |
XM_017000819.1 | XP_016856308.1 | 71 - 72 | 10077 | 73 | |
XM_017000820.1 | XP_016856309.1 | 70 - 71 | 10059 | 73 | |
XM_017000821.1 | XP_016856310.1 | 67 - 68 | 9751 | 73 | |
GenBank mRNA | AB007936.2 | - | 55 - 56 | 7850 | 73 |
AK001249.2 | - | 4 - 5 | 809 | 73 | |
AK027078.1 | - | 10 - 11 | 1543 | 73 | |
AK126972.1 | - | 8 - 9 | 1900 | 73 | |
AL117402.1 | - | 4 - 5 | 792 | 73 | |
BC041069.1 | - | 11 - 12 | 1679 | 73 | |
BC052802.1 | - | 12 - 13 | 1693 | 73 | |
BC082968.1 | - | 12 - 13 | 1698 | 73 | |
BC151232.1 | - | 55 - 56 | 7850 | 73 |