The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005117.2 | NP_005108.1 | 1 - 2 | 690 | 99 |
GenBank mRNA | AB018122.1 | - | 1 - 2 | 227 | 99 |
AF110400.1 | - | 1 - 2 | 690 | 99 | |
AY358302.1 | - | 1 - 2 | 690 | 99 | |
BC017664.1 | - | 1 - 2 | 358 | 99 | |
BT006729.1 | - | 1 - 2 | 227 | 99 |