The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015205.2 | NP_056020.2 | 20 - 21 | 2506 | 119 |
NM_032189.3 | NP_115565.3 | 20 - 21 | 2506 | 119 | |
XM_005268299.4 | XP_005268356.1 | 20 - 21 | 2801 | 119 | |
XM_005268300.4 | XP_005268357.1 | 20 - 21 | 2800 | 119 | |
XM_005268301.4 | XP_005268358.1 | 20 - 21 | 2801 | 119 | |
XM_005268303.4 | XP_005268360.1 | 20 - 21 | 2801 | 119 | |
XM_005268305.4 | XP_005268362.1 | 20 - 21 | 2800 | 119 | |
XM_005268306.4 | XP_005268363.1 | 20 - 21 | 2800 | 119 | |
XM_011537480.2 | XP_011535782.1 | 20 - 21 | 2801 | 119 | |
XM_017020490.1 | XP_016875979.1 | 20 - 21 | 2801 | 119 | |
XM_017020491.1 | XP_016875980.1 | 20 - 21 | 2801 | 119 | |
XM_017020492.1 | XP_016875981.1 | 20 - 21 | 2800 | 119 | |
GenBank mRNA | AB028944.2 | - | 19 - 20 | 2323 | 119 |
AK128241.1 | - | 19 - 20 | 2369 | 119 | |
AL161996.1 | - | 4 - 5 | 680 | 119 | |
BC024154.2 | - | 19 - 20 | 2342 | 119 |