The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020728.2 | NP_065779.1 | 1 - 2 | 544 | 102 |
GenBank mRNA | AB033054.2 | - | 2 - 3 | 330 | 102 |
AK308824.1 | - | 1 - 2 | 152 | 102 | |
AL833233.1 | - | 1 - 2 | 153 | 102 | |
AY368150.1 | - | 1 - 2 | 544 | 102 | |
DQ993201.1 | - | 3 - 4 | 562 | 102 |