The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247996.1 | NP_001234925.1 | 28 - 29 | 3033 | 75 |
NM_018482.3 | NP_060952.2 | 27 - 28 | 2931 | 75 | |
XM_005250925.1 | XP_005250982.1 | 28 - 29 | 2940 | 75 | |
XM_006716563.3 | XP_006716626.1 | 28 - 29 | 2759 | 75 | |
XM_006716564.1 | XP_006716627.1 | 29 - 30 | 2918 | 75 | |
XM_006716565.3 | XP_006716628.1 | 27 - 28 | 2755 | 75 | |
XM_006716566.1 | XP_006716629.1 | 27 - 28 | 2769 | 75 | |
XM_011517052.2 | XP_011515354.1 | 28 - 29 | 2756 | 75 | |
XM_011517053.1 | XP_011515355.1 | 28 - 29 | 2962 | 75 | |
XM_017013467.1 | XP_016868956.1 | 26 - 27 | 2777 | 75 | |
XM_017013468.1 | XP_016868957.1 | 27 - 28 | 3001 | 75 | |
XM_017013469.1 | XP_016868958.1 | 26 - 27 | 2760 | 75 | |
GenBank mRNA | AB033075.1 | - | 21 - 22 | 2164 | 75 |
BC137135.1 | - | 26 - 27 | 2728 | 75 | |
BC144493.1 | - | 27 - 28 | 2830 | 75 | |
BX537768.1 | - | 20 - 21 | 2184 | 75 |