The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006657.1 | NP_001006658.1 | 15 - 16 | 1663 | 126 |
NM_020779.3 | NP_065830.2 | 14 - 15 | 1630 | 126 | |
XM_011533007.2 | XP_011531309.1 | 4 - 5 | 355 | 126 | |
GenBank mRNA | AB037757.2 | - | 15 - 16 | 1671 | 126 |
AK091205.1 | - | 8 - 9 | 998 | 126 | |
AK122917.1 | - | 14 - 15 | 1555 | 126 | |
AK315094.1 | - | 14 - 15 | 1555 | 126 | |
AL832903.1 | - | 3 - 4 | 160 | 126 | |
BC036659.2 | - | 14 - 15 | 1629 | 126 |