The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285439.1 | NP_001272368.1 | 17 - 18 | 1600 | 94 |
NM_001285440.1 | NP_001272369.1 | 17 - 18 | 1598 | 94 | |
NM_152756.4 | NP_689969.2 | 17 - 18 | 1600 | 94 | |
XM_006714463.3 | XP_006714526.1 | 17 - 18 | 1613 | 94 | |
XM_011514005.2 | XP_011512307.1 | 17 - 18 | 1613 | 94 | |
XM_011514006.2 | XP_011512308.1 | 16 - 17 | 1417 | 94 | |
XM_017009311.1 | XP_016864800.1 | 16 - 17 | 1533 | 94 | |
XM_017009312.1 | XP_016864801.1 | 16 - 17 | 1533 | 94 | |
XM_017009313.1 | XP_016864802.1 | 16 - 17 | 1554 | 94 | |
XM_017009314.1 | XP_016864803.1 | 16 - 17 | 1523 | 94 | |
XM_017009315.1 | XP_016864804.1 | 16 - 17 | 1523 | 94 | |
GenBank mRNA | AB082530.1 | - | 2 - 3 | 271 | 94 |
AY515854.1 | - | 17 - 18 | 1596 | 94 | |
BC137163.1 | - | 17 - 18 | 1593 | 94 | |
BC137164.1 | - | 17 - 18 | 1593 | 94 | |
BC144509.1 | - | 17 - 18 | 1593 | 94 | |
CR749280.1 | - | 17 - 18 | 1579 | 94 |