Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBFOX1
Entrez Gene ID
54715
Gene Name
RNA binding protein, fox-1 homolog 1
Gene Aliases
2BP1, A2BP1, FOX-1, FOX1, HRNBP1
Chromosome Location
Chr.16: 5239749 - 7713343 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142333.1 NP_001135805.1 5 - 6 1261 90
NM_001142334.1 NP_001135806.1 3 - 4 493 90
NM_001308117.1 NP_001295046.1 5 - 6 598 90
NM_018723.3 NP_061193.2 5 - 6 1261 90
NM_145891.2 NP_665898.1 2 - 3 585 90
NM_145892.2 NP_665899.1 2 - 3 585 90
NM_145893.2 NP_665900.1 2 - 3 585 90
XM_005255386.3 XP_005255443.1 2 - 3 511 90
XM_005255387.3 XP_005255444.1 2 - 3 490 90
XM_005255388.4 XP_005255445.1 2 - 3 546 90
XM_005255390.3 XP_005255447.1 5 - 6 1383 90
XM_005255391.3 XP_005255448.1 4 - 5 393 90
XM_005255394.4 XP_005255451.1 2 - 3 490 90
XM_011522546.2 XP_011520848.1 2 - 3 490 90
XM_011522547.2 XP_011520849.1 2 - 3 490 90
XM_011522548.2 XP_011520850.1 2 - 3 490 90
XM_017023318.1 XP_016878807.1 6 - 7 888 90
XM_017023319.1 XP_016878808.1 8 - 9 884 90
XM_017023320.1 XP_016878809.1 5 - 6 1292 90
XM_017023321.1 XP_016878810.1 4 - 5 2354 90
XM_017023322.1 XP_016878811.1 4 - 5 2354 90
XM_017023323.1 XP_016878812.1 2 - 3 511 90
XM_017023324.1 XP_016878813.1 5 - 6 1292 90
XM_017023325.1 XP_016878814.1 3 - 4 890 90
XM_017023326.1 XP_016878815.1 5 - 6 1383 90
XM_017023327.1 XP_016878816.1 2 - 3 511 90
XM_017023328.1 XP_016878817.1 5 - 6 1292 90
XM_017023329.1 XP_016878818.1 5 - 6 1292 90
XM_017023330.1 XP_016878819.1 4 - 5 1831 90
XM_017023331.1 XP_016878820.1 2 - 3 2297 90
XM_017023332.1 XP_016878821.1 2 - 3 511 90
XM_017023333.1 XP_016878822.1 4 - 5 393 90
XM_017023334.1 XP_016878823.1 2 - 3 511 90
XM_017023335.1 XP_016878824.1 5 - 6 1383 90
XM_017023336.1 XP_016878825.1 4 - 5 393 90
XM_017023337.1 XP_016878826.1 2 - 3 511 90
XM_017023338.1 XP_016878827.1 4 - 5 393 90
XM_017023339.1 XP_016878828.1 2 - 3 537 90
XM_017023340.1 XP_016878829.1 4 - 5 393 90
XM_017023341.1 XP_016878830.1 5 - 6 1383 90
XM_017023342.1 XP_016878831.1 4 - 5 393 90
XM_017023343.1 XP_016878832.1 2 - 3 540 90
GenBank mRNA AF094849.1 - 2 - 3 586 90
AF107203.1 - 5 - 6 1259 90
AF109106.1 - 2 - 3 586 90
AF229057.1 - 2 - 3 588 90
AF448859.1 - 5 - 6 1259 90
AK293951.1 - 5 - 6 598 90
AL713700.1 - 4 - 5 1108 90
BC113691.1 - 2 - 3 379 90
BC143816.1 - 2 - 3 379 90
KJ534926.1 - 2 - 3 333 90
KJ535000.1 - 2 - 3 333 90
KJ535001.1 - 2 - 3 333 90

View Full Product Details