The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032172.2 | NP_115548.1 | 11 - 12 | 1351 | 76 |
XM_005249883.4 | XP_005249940.1 | 11 - 12 | 1298 | 76 | |
XM_006715790.1 | XP_006715853.1 | 11 - 12 | 1298 | 76 | |
XM_006715791.2 | XP_006715854.1 | 14 - 15 | 1804 | 76 | |
XM_011515573.1 | XP_011513875.1 | 15 - 16 | 1863 | 76 | |
XM_011515574.1 | XP_011513876.1 | 12 - 13 | 1387 | 76 | |
XM_011515577.1 | XP_011513879.1 | 10 - 11 | 1016 | 76 | |
XM_011515578.1 | XP_011513880.1 | 12 - 13 | 1438 | 76 | |
GenBank mRNA | AJ601395.1 | - | 10 - 11 | 1228 | 76 |
AK022759.1 | - | 11 - 12 | 1295 | 76 | |
AK301527.1 | - | 9 - 10 | 823 | 76 | |
AK309375.1 | - | 10 - 11 | 1092 | 76 | |
AY618868.1 | - | 11 - 12 | 1295 | 76 | |
BC060846.1 | - | 11 - 12 | 1290 | 76 | |
BC132862.1 | - | 10 - 11 | 1248 | 76 | |
BC136656.1 | - | 9 - 10 | 2181 | 76 |