Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLITRK5
Entrez Gene ID
26050
Gene Name
SLIT and NTRK like family member 5
Gene Aliases
LRRC11, bA364G4.2
Chromosome Location
Chr.13: 87671328 - 87679619 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015567.1 NP_056382.1 1 - 2 205 96
GenBank mRNA AK024251.1 - 1 - 2 205 96
BC098106.1 - 1 - 2 113 96
BC098287.1 - 1 - 2 113 96
BC103509.1 - 1 - 2 113 96

View Full Product Details