Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS13C
Entrez Gene ID
54832
Gene Name
vacuolar protein sorting 13 homolog C
Gene Aliases
PARK23
Chromosome Location
Chr.15: 61852389 - 62060465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018088.2 NP_001018098.1 16 - 17 1442 116
NM_017684.4 NP_060154.3 14 - 15 1313 116
NM_018080.3 NP_060550.2 14 - 15 1313 116
NM_020821.2 NP_065872.1 16 - 17 1442 116
XM_011521713.2 XP_011520015.1 16 - 17 1403 116
XM_011521714.2 XP_011520016.1 16 - 17 1403 116
GenBank mRNA AB037842.1 - 16 - 17 1425 116
AJ608770.1 - 14 - 15 1296 116
AJ608771.1 - 16 - 17 1425 116
AJ626860.1 - 14 - 15 1296 116
AJ626861.1 - 16 - 17 1425 116
AK025364.1 - 11 - 12 964 116

View Full Product Details