Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SASH1
Entrez Gene ID
23328
Gene Name
SAM and SH3 domain containing 1
Gene Aliases
SH3D6A, dJ323M4, dJ323M4.1
Chromosome Location
Chr.6: 148193440 - 148552049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015278.3 NP_056093.3 7 - 8 1098 87
XM_011535640.1 XP_011533942.1 7 - 8 541 87
XM_017010598.1 XP_016866087.1 7 - 8 1330 87
XM_017010599.1 XP_016866088.1 7 - 8 653 87
XM_017010600.1 XP_016866089.1 7 - 8 541 87
XM_017010601.1 XP_016866090.1 6 - 7 755 87
XM_017010602.1 XP_016866091.1 5 - 6 452 87
XM_017010603.1 XP_016866092.1 5 - 6 391 87
XM_017010604.1 XP_016866093.1 4 - 5 300 87
XM_017010605.1 XP_016866094.1 4 - 5 266 87
XM_017010607.1 XP_016866096.1 8 - 9 721 87
GenBank mRNA AB018333.1 - 7 - 8 841 87
AJ507735.1 - 7 - 8 629 87
AK024403.1 - 1 - 2 83 87
AK025495.1 - 7 - 8 1098 87

View Full Product Details