The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173515.2 | NP_775786.2 | 3 - 4 | 923 | 127 |
XM_011535485.2 | XP_011533787.1 | 3 - 4 | 3406 | 127 | |
GenBank mRNA | AK055911.1 | - | 3 - 4 | 963 | 127 |
AK309680.1 | - | 3 - 4 | 529 | 127 | |
AY328891.1 | - | 3 - 4 | 923 | 127 | |
AY328892.1 | - | 3 - 4 | 923 | 127 | |
AY328893.1 | - | 3 - 4 | 923 | 127 | |
BC060761.1 | - | 3 - 4 | 845 | 127 |