Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41610101-41610601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT16
Entrez Gene ID
3868
Gene Name
keratin 16
Gene Aliases
CK16, FNEPPK, K16, K1CP, KRT16A, NEPPK, PC1
UniGene
Hs.655160
Gene Chromosome Location
Chr.17: 41609778-41612827 on Build GRCh38
Transcripts
NM_005557.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146499 c.1108A>T p.M370L Chr.17: 41610503-41610503 on Build GRCh38
4917632 c.1073A>G p.E358G Chr.17: 41610538-41610538 on Build GRCh38
245337 c.1267G>A p.G423S Chr.17: 41610344-41610344 on Build GRCh38
6898894 c.1290C>T p.S430S Chr.17: 41610227-41610227 on Build GRCh38
5016568 c.1210_1214delCTGGA p.L404fs*>69 Chr.17: 41610397-41610401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755660186 Chr.17: 41610560-41610563 on Build GRCh38
rs771720525 Chr.17: 41610500-41610500 on Build GRCh38
rs748198038 Chr.17: 41610283-41610283 on Build GRCh38
rs754961471 Chr.17: 41610479-41610479 on Build GRCh38
rs752290870 Chr.17: 41610560-41610561 on Build GRCh38
rs59328451 Chr.17: 41610549-41610549 on Build GRCh38
rs568310923 Chr.17: 41610295-41610295 on Build GRCh38
rs751535304 Chr.17: 41610256-41610256 on Build GRCh38
rs758322165 Chr.17: 41610168-41610168 on Build GRCh38
rs749633528 Chr.17: 41610579-41610579 on Build GRCh38
rs773801621 Chr.17: 41610196-41610196 on Build GRCh38
rs751361060 Chr.17: 41610134-41610134 on Build GRCh38
rs558871243 Chr.17: 41610124-41610124 on Build GRCh38
rs112929229 Chr.17: 41610237-41610237 on Build GRCh38
rs757273464 Chr.17: 41610548-41610548 on Build GRCh38
rs762324547 Chr.17: 41610312-41610312 on Build GRCh38
rs751249671 Chr.17: 41610319-41610319 on Build GRCh38
rs780464132 Chr.17: 41610461-41610461 on Build GRCh38
rs751757007 Chr.17: 41610547-41610547 on Build GRCh38
rs746390259 Chr.17: 41610450-41610450 on Build GRCh38
rs745725822 Chr.17: 41610270-41610270 on Build GRCh38
rs780475822 Chr.17: 41610569-41610569 on Build GRCh38
rs753131774 Chr.17: 41610175-41610175 on Build GRCh38
rs148365727 Chr.17: 41610386-41610386 on Build GRCh38
rs751388606 Chr.17: 41610341-41610341 on Build GRCh38
rs772428048 Chr.17: 41610195-41610195 on Build GRCh38
rs768261124 Chr.17: 41610395-41610395 on Build GRCh38
rs763894959 Chr.17: 41610254-41610254 on Build GRCh38
rs765910106 Chr.17: 41610157-41610157 on Build GRCh38
rs372742922 Chr.17: 41610123-41610123 on Build GRCh38
rs757745536 Chr.17: 41610381-41610381 on Build GRCh38
rs759285850 Chr.17: 41610525-41610525 on Build GRCh38
rs762413188 Chr.17: 41610141-41610141 on Build GRCh38
rs777902557 Chr.17: 41610439-41610439 on Build GRCh38
rs773915759 Chr.17: 41610475-41610475 on Build GRCh38
rs749532028 Chr.17: 41610183-41610183 on Build GRCh38
rs776202401 Chr.17: 41610223-41610223 on Build GRCh38
rs762797152 Chr.17: 41610282-41610283 on Build GRCh38
rs533232973 Chr.17: 41610178-41610178 on Build GRCh38
rs111470587 Chr.17: 41610238-41610238 on Build GRCh38
rs760487188 Chr.17: 41610414-41610414 on Build GRCh38
rs773899833 Chr.17: 41610399-41610399 on Build GRCh38
rs750570209 Chr.17: 41610561-41610561 on Build GRCh38
rs768135950 Chr.17: 41610142-41610142 on Build GRCh38
rs367591548 Chr.17: 41610194-41610194 on Build GRCh38
rs934666354 Chr.17: 41610326-41610326 on Build GRCh38
rs761337135 Chr.17: 41610482-41610482 on Build GRCh38
rs527635390 Chr.17: 41610279-41610279 on Build GRCh38
rs889918210 Chr.17: 41610411-41610411 on Build GRCh38
rs781137610 Chr.17: 41610266-41610266 on Build GRCh38
rs138335461 Chr.17: 41610356-41610356 on Build GRCh38
rs768333887 Chr.17: 41610474-41610474 on Build GRCh38
rs761807016 Chr.17: 41610342-41610342 on Build GRCh38
rs760683390 Chr.17: 41610512-41610512 on Build GRCh38
rs760901329 Chr.17: 41610204-41610204 on Build GRCh38
rs565842111 Chr.17: 41610287-41610287 on Build GRCh38
rs373241507 Chr.17: 41610458-41610458 on Build GRCh38
rs766782137 Chr.17: 41610213-41610213 on Build GRCh38
rs201334428 Chr.17: 41610503-41610503 on Build GRCh38
rs267607407 Chr.17: 41610341-41610341 on Build GRCh38
rs386797103 Chr.17: 41610252-41610253 on Build GRCh38
rs750221600 Chr.17: 41610347-41610347 on Build GRCh38
rs150916635 Chr.17: 41610514-41610514 on Build GRCh38
rs746746409 Chr.17: 41610169-41610169 on Build GRCh38
rs377349355 Chr.17: 41610442-41610442 on Build GRCh38
rs375579387 Chr.17: 41610162-41610162 on Build GRCh38
rs761278361 Chr.17: 41610400-41610400 on Build GRCh38
rs374761160 Chr.17: 41610158-41610158 on Build GRCh38
rs777901534 Chr.17: 41610358-41610358 on Build GRCh38
rs71357510 Chr.17: 41610134-41610135 on Build GRCh38
rs754709110 Chr.17: 41610507-41610507 on Build GRCh38
rs749674424 Chr.17: 41610464-41610464 on Build GRCh38
rs141569849 Chr.17: 41610443-41610443 on Build GRCh38
rs755238938 Chr.17: 41610271-41610271 on Build GRCh38
rs766279027 Chr.17: 41610427-41610427 on Build GRCh38
rs781631228 Chr.17: 41610449-41610449 on Build GRCh38
rs774567335 Chr.17: 41610193-41610193 on Build GRCh38
rs762673256 Chr.17: 41610388-41610388 on Build GRCh38
rs372739869 Chr.17: 41610515-41610515 on Build GRCh38
rs544761071 Chr.17: 41610138-41610138 on Build GRCh38
rs574276845 Chr.17: 41610130-41610130 on Build GRCh38
rs766195354 Chr.17: 41610311-41610338 on Build GRCh38
rs150453468 Chr.17: 41610300-41610300 on Build GRCh38
rs771443776 Chr.17: 41610369-41610369 on Build GRCh38
rs56259134 Chr.17: 41610359-41610359 on Build GRCh38
rs756004119 Chr.17: 41610350-41610350 on Build GRCh38
rs763676345 Chr.17: 41610323-41610323 on Build GRCh38
rs969859566 Chr.17: 41610554-41610554 on Build GRCh38
rs750731967 Chr.17: 41610150-41610150 on Build GRCh38
rs549128698 Chr.17: 41610289-41610289 on Build GRCh38
rs376747202 Chr.17: 41610459-41610459 on Build GRCh38
rs756941943 Chr.17: 41610265-41610265 on Build GRCh38
rs757104566 Chr.17: 41610172-41610172 on Build GRCh38
rs768788608 Chr.17: 41610188-41610188 on Build GRCh38
rs761142594 Chr.17: 41610152-41610152 on Build GRCh38
rs373125552 Chr.17: 41610410-41610410 on Build GRCh38
rs560220901 Chr.17: 41610253-41610253 on Build GRCh38
rs752273996 Chr.17: 41610167-41610167 on Build GRCh38
rs138288025 Chr.17: 41610355-41610355 on Build GRCh38
rs375085095 Chr.17: 41610540-41610540 on Build GRCh38
rs764733739 Chr.17: 41610165-41610165 on Build GRCh38
rs7406899 Chr.17: 41610252-41610252 on Build GRCh38
rs200457676 Chr.17: 41610568-41610568 on Build GRCh38
rs535973339 Chr.17: 41610140-41610140 on Build GRCh38
rs369631543 Chr.17: 41610233-41610233 on Build GRCh38
rs777623129 Chr.17: 41610569-41610570 on Build GRCh38
rs759188113 Chr.17: 41610231-41610231 on Build GRCh38
rs370949361 Chr.17: 41610284-41610284 on Build GRCh38
rs775068482 Chr.17: 41610387-41610387 on Build GRCh38
rs780946255 Chr.17: 41610481-41610482 on Build GRCh38
rs753640770 Chr.17: 41610428-41610428 on Build GRCh38
rs377221209 Chr.17: 41610257-41610257 on Build GRCh38
rs368945986 Chr.17: 41610385-41610385 on Build GRCh38
rs759938445 Chr.17: 41610291-41610291 on Build GRCh38

View Full Product Details