Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC27
Entrez Gene ID
148870
Gene Name
coiled-coil domain containing 27
Gene Aliases
-
Chromosome Location
Chr.1: 3752398 - 3771645 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152492.2 NP_689705.2 2 - 3 522 54
XM_011540780.1 XP_011539082.1 2 - 3 525 54
XM_017000402.1 XP_016855891.1 2 - 3 525 54
XM_017000403.1 XP_016855892.1 2 - 3 525 54
XM_017000404.1 XP_016855893.1 2 - 3 525 54
XM_017000405.1 XP_016855894.1 2 - 3 525 54
XM_017000406.1 XP_016855895.1 3 - 4 339 54
GenBank mRNA AK057387.1 - 2 - 3 525 54
BC112108.1 - 2 - 3 523 54
BC112112.1 - 2 - 3 522 54
DB048639.1 - 2 - 3 480 54

View Full Product Details