Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099280.2 NP_001092750.1 4 - 5 407 118
NM_001099281.2 NP_001092751.1 3 - 4 287 118
NM_001288814.1 NP_001275743.1 3 - 4 294 118
NM_032450.2 NP_115826.2 4 - 5 407 118
XM_011517270.2 XP_011515572.1 4 - 5 545 118
XM_011517271.2 XP_011515573.1 4 - 5 544 118
XM_011517272.2 XP_011515574.1 4 - 5 545 118
XM_011517273.2 XP_011515575.1 4 - 5 543 118
XM_011517274.2 XP_011515576.1 4 - 5 543 118
XM_011517275.2 XP_011515577.1 4 - 5 543 118
XM_011517276.2 XP_011515578.1 4 - 5 571 118
XM_011517277.1 XP_011515579.1 4 - 5 407 118
XM_011517278.1 XP_011515580.1 3 - 4 287 118
XM_011517279.2 XP_011515581.1 3 - 4 451 118
XM_011517283.2 XP_011515585.1 4 - 5 366 118
XM_011517285.1 XP_011515587.1 3 - 4 287 118
XM_011517286.2 XP_011515588.1 4 - 5 539 118
XM_017013802.1 XP_016869291.1 4 - 5 543 118
XM_017013803.1 XP_016869292.1 4 - 5 542 118
XM_017013804.1 XP_016869293.1 4 - 5 407 118
XM_017013805.1 XP_016869294.1 3 - 4 287 118
XM_017013807.1 XP_016869296.1 4 - 5 540 118
GenBank mRNA AB058736.1 - 3 - 4 299 118
AK093563.1 - 4 - 5 406 118
AK295740.1 - 3 - 4 287 118
AL834309.1 - 3 - 4 291 118
BC057824.1 - 4 - 5 387 118
BC067855.1 - 4 - 5 387 118
BC092486.1 - 3 - 4 267 118
DA934047.1 - 4 - 5 407 118

View Full Product Details