Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMD14
Entrez Gene ID
201191
Gene Name
sterile alpha motif domain containing 14
Gene Aliases
-
Chromosome Location
Chr.17: 50111309 - 50129882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257359.1 NP_001244288.1 8 - 9 1319 124
NM_174920.3 NP_777580.1 9 - 10 1403 124
XM_011524490.2 XP_011522792.1 5 - 6 505 124
XM_017024322.1 XP_016879811.1 7 - 8 1703 124
XM_017024323.1 XP_016879812.1 9 - 10 1486 124
XM_017024324.1 XP_016879813.1 9 - 10 2258 124
GenBank mRNA AF035306.1 - 2 - 3 191 124
AK094209.1 - 8 - 9 1250 124
AK095809.1 - 5 - 6 620 124
AK225767.1 - 8 - 9 1250 124
AK291482.1 - 8 - 9 1246 124
AY148482.1 - 9 - 10 1324 124
BC029872.1 - 4 - 5 431 124
BC047081.1 - 7 - 8 973 124
BC136485.1 - 8 - 9 1232 124
BC141822.1 - 8 - 9 1259 124
BC142728.1 - 8 - 9 1216 124

View Full Product Details