The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001370.1 | NP_001361.1 | 8 - 9 | 1455 | 89 |
XM_006711956.2 | XP_006712019.1 | 8 - 9 | 1560 | 89 | |
XM_011532649.2 | XP_011530951.1 | 8 - 9 | 1409 | 89 | |
XM_011532650.2 | XP_011530952.1 | 8 - 9 | 1410 | 89 | |
XM_011532652.1 | XP_011530954.1 | 8 - 9 | 1455 | 89 | |
XM_011532653.1 | XP_011530955.1 | 8 - 9 | 1455 | 89 | |
XM_011532654.1 | XP_011530956.1 | 7 - 8 | 1188 | 89 | |
XM_011532655.2 | XP_011530957.1 | 4 - 5 | 477 | 89 | |
XM_011532657.1 | XP_011530959.1 | 2 - 3 | 216 | 89 | |
XM_011532660.1 | XP_011530962.1 | 8 - 9 | 1455 | 89 | |
XM_017003521.1 | XP_016859010.1 | 8 - 9 | 1455 | 89 | |
XM_017003522.1 | XP_016859011.1 | 3 - 4 | 1281 | 89 | |
XM_017003523.1 | XP_016859012.1 | 3 - 4 | 448 | 89 | |
XM_017003524.1 | XP_016859013.1 | 8 - 9 | 1455 | 89 | |
GenBank mRNA | AK094676.1 | - | 4 - 5 | 668 | 89 |
BC104884.1 | - | 2 - 3 | 361 | 89 | |
BC113424.1 | - | 2 - 3 | 361 | 89 | |
BC143666.1 | - | 2 - 3 | 361 | 89 |