The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286152.1 | NP_001273081.1 | 5 - 6 | 723 | 115 |
NM_198275.2 | NP_938016.1 | 5 - 6 | 759 | 115 | |
NR_104404.1 | 2 - 3 | 215 | 115 | ||
NR_104405.1 | 4 - 5 | 548 | 115 | ||
XM_006718775.2 | XP_006718838.1 | 5 - 6 | 1111 | 115 | |
XM_017017304.1 | XP_016872793.1 | 5 - 6 | 870 | 115 | |
GenBank mRNA | AK095399.1 | - | 5 - 6 | 711 | 115 |
AK289390.1 | - | 5 - 6 | 711 | 115 | |
AK296951.1 | - | 4 - 5 | 499 | 115 | |
AK304292.1 | - | 5 - 6 | 702 | 115 | |
AK310966.1 | - | 2 - 3 | 194 | 115 | |
AY358626.1 | - | 5 - 6 | 687 | 115 | |
BC113586.1 | - | 5 - 6 | 741 | 115 | |
BC143897.1 | - | 5 - 6 | 705 | 115 |