The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173854.5 | NP_776253.3 | 8 - 9 | 2109 | 61 |
XM_005245069.2 | XP_005245126.1 | 8 - 9 | 1785 | 61 | |
GenBank mRNA | AJ514402.1 | - | 7 - 8 | 1196 | 61 |
AK125075.1 | - | 7 - 8 | 870 | 61 | |
AK128069.1 | - | 7 - 8 | 1362 | 61 | |
AK314732.1 | - | 7 - 8 | 1267 | 61 | |
AL831974.1 | - | 8 - 9 | 1945 | 61 | |
AL832001.1 | - | 8 - 9 | 1945 | 61 | |
AL832362.1 | - | 8 - 9 | 1945 | 61 | |
BC136303.1 | - | 7 - 8 | 1590 | 61 | |
BX648979.1 | - | 8 - 9 | 1983 | 61 |