The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011649.2 | NP_001011649.1 | 14 - 15 | 1812 | 103 |
NM_001272039.1 | NP_001258968.1 | 14 - 15 | 1812 | 103 | |
NM_018249.5 | NP_060719.4 | 14 - 15 | 1812 | 103 | |
NR_073554.1 | 14 - 15 | 1812 | 103 | ||
NR_073555.1 | 14 - 15 | 1812 | 103 | ||
NR_073556.1 | 15 - 16 | 1939 | 103 | ||
NR_073557.1 | 14 - 15 | 1812 | 103 | ||
NR_073558.1 | 14 - 15 | 1809 | 103 | ||
XM_006717182.1 | XP_006717245.1 | 14 - 15 | 1801 | 103 | |
XM_006717185.1 | XP_006717248.1 | 14 - 15 | 1801 | 103 | |
XM_011518860.1 | XP_011517162.1 | 14 - 15 | 1801 | 103 | |
XM_017014921.1 | XP_016870410.1 | 14 - 15 | 1801 | 103 | |
XM_017014922.1 | XP_016870411.1 | 8 - 9 | 1030 | 103 | |
XM_017014923.1 | XP_016870412.1 | 14 - 15 | 1801 | 103 | |
XM_017014924.1 | XP_016870413.1 | 5 - 6 | 706 | 103 | |
GenBank mRNA | AB046853.2 | - | 14 - 15 | 1781 | 103 |
AF448860.1 | - | 14 - 15 | 1762 | 103 | |
AK122913.1 | - | 14 - 15 | 1801 | 103 | |
AL833481.1 | - | 14 - 15 | 1800 | 103 | |
BC136275.1 | - | 14 - 15 | 1647 | 103 | |
BC143762.1 | - | 14 - 15 | 1647 | 103 | |
BC146782.1 | - | 14 - 15 | 1781 | 103 | |
BX537421.1 | - | 14 - 15 | 1797 | 103 | |
BX537708.1 | - | 14 - 15 | 1801 | 103 | |
BX537759.1 | - | 14 - 15 | 1800 | 103 | |
BX640896.1 | - | 8 - 9 | 1024 | 103 | |
CR936622.1 | - | 7 - 8 | 942 | 103 | |
CR936717.1 | - | 7 - 8 | 942 | 103 | |
KP294527.1 | - | 14 - 15 | 1812 | 103 |