The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001265612.1 | NP_001252541.1 | 29 - 30 | 4323 | 61 |
NM_016284.4 | NP_057368.3 | 29 - 30 | 4338 | 61 | |
NM_206999.2 | NP_996882.1 | 29 - 30 | 4338 | 61 | |
NR_049763.1 | 29 - 30 | 4323 | 61 | ||
GenBank mRNA | AB023224.2 | - | 16 - 17 | 2383 | 61 |
AK302495.1 | - | 6 - 7 | 708 | 61 | |
AL117492.1 | - | 1 - 2 | 177 | 61 | |
AL832769.1 | - | 29 - 30 | 4287 | 61 | |
AL833549.1 | - | 29 - 30 | 4297 | 61 | |
BC024317.1 | - | 12 - 13 | 1737 | 61 | |
BC040523.1 | - | 29 - 30 | 4290 | 61 | |
BX537840.1 | - | 29 - 30 | 4269 | 61 | |
BX647815.1 | - | 29 - 30 | 4278 | 61 | |
CR749237.1 | - | 22 - 23 | 3278 | 61 | |
EF553522.1 | - | 29 - 30 | 4272 | 61 |