The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198578.3 | NP_940980.3 | 47 - 48 | 7149 | 108 |
XM_005268629.3 | XP_005268686.1 | 47 - 48 | 7265 | 108 | |
XM_011537877.2 | XP_011536179.1 | 47 - 48 | 7265 | 108 | |
XM_017018787.1 | XP_016874276.1 | 26 - 27 | 4093 | 108 | |
XM_017018788.1 | XP_016874277.1 | 22 - 23 | 3337 | 108 | |
GenBank mRNA | AK122640.1 | - | 14 - 15 | 2057 | 108 |
AK131537.1 | - | 22 - 23 | 3453 | 108 | |
AL832453.2 | - | 26 - 27 | 4322 | 108 | |
AL834529.1 | - | 4 - 5 | 647 | 108 | |
AY792511.1 | - | 47 - 48 | 7028 | 108 | |
BC117180.1 | - | 47 - 48 | 7086 | 108 |