The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015150.1 | NP_055965.1 | 9 - 10 | 1616 | 107 |
XM_005264985.1 | XP_005265042.1 | 9 - 10 | 1573 | 107 | |
XM_005264986.2 | XP_005265043.1 | 9 - 10 | 1470 | 107 | |
XM_006713069.1 | XP_006713132.1 | 9 - 10 | 1479 | 107 | |
XM_011533529.1 | XP_011531831.1 | 11 - 12 | 2013 | 107 | |
XM_011533530.1 | XP_011531832.1 | 9 - 10 | 1641 | 107 | |
XM_011533531.1 | XP_011531833.1 | 10 - 11 | 1897 | 107 | |
XM_017006007.1 | XP_016861496.1 | 8 - 9 | 1484 | 107 | |
GenBank mRNA | AK074446.1 | - | 9 - 10 | 1422 | 107 |
AK301024.1 | - | 8 - 9 | 1309 | 107 | |
AK303846.1 | - | 5 - 6 | 613 | 107 | |
AK310258.1 | - | 3 - 4 | 317 | 107 | |
AY237654.1 | - | 9 - 10 | 1607 | 107 | |
BC006400.2 | - | 5 - 6 | 544 | 107 | |
BC014032.2 | - | 5 - 6 | 544 | 107 | |
BC032349.1 | - | 8 - 9 | 1289 | 107 | |
BC051336.1 | - | 9 - 10 | 1393 | 107 | |
BC069209.1 | - | 9 - 10 | 1393 | 107 | |
BC098270.1 | - | 9 - 10 | 1440 | 107 | |
BC100667.1 | - | 9 - 10 | 1440 | 107 | |
BC100669.1 | - | 9 - 10 | 1440 | 107 | |
BC121795.1 | - | 9 - 10 | 1440 | 107 | |
D42043.1 | - | 9 - 10 | 1544 | 107 |