Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27560755-27561339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf72
Entrez Gene ID
203228
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
UniGene
Hs.493639
Gene Chromosome Location
Chr.9: 27546546-27573866 on Build GRCh38
Transcripts
NM_001256054.2 NM_018325.4 NM_145005.6

SNP Detail

SNP ID Chromosome Location
rs1029561480 Chr.9: 27561244-27561244 on Build GRCh38
rs567070216 Chr.9: 27560800-27560800 on Build GRCh38
rs933508836 Chr.9: 27561173-27561173 on Build GRCh38
rs538585248 Chr.9: 27561127-27561127 on Build GRCh38
rs955288267 Chr.9: 27561259-27561259 on Build GRCh38
rs922521465 Chr.9: 27560937-27560937 on Build GRCh38
rs878966701 Chr.9: 27560809-27560809 on Build GRCh38
rs780646871 Chr.9: 27561050-27561050 on Build GRCh38
rs981925831 Chr.9: 27561270-27561270 on Build GRCh38
rs1052719316 Chr.9: 27560812-27560812 on Build GRCh38
rs955319198 Chr.9: 27560946-27560946 on Build GRCh38
rs1005368758 Chr.9: 27560883-27560883 on Build GRCh38
rs888176698 Chr.9: 27560858-27560858 on Build GRCh38
rs997685909 Chr.9: 27560910-27560910 on Build GRCh38
rs564985448 Chr.9: 27561168-27561168 on Build GRCh38
rs533995553 Chr.9: 27560801-27560801 on Build GRCh38
rs149398015 Chr.9: 27561124-27561124 on Build GRCh38
rs1013885466 Chr.9: 27560839-27560839 on Build GRCh38
rs748306604 Chr.9: 27561061-27561061 on Build GRCh38
rs747539741 Chr.9: 27560944-27560944 on Build GRCh38
rs896747555 Chr.9: 27561128-27561128 on Build GRCh38
rs935578989 Chr.9: 27561103-27561103 on Build GRCh38
rs1039431665 Chr.9: 27561160-27561160 on Build GRCh38
rs893808117 Chr.9: 27561242-27561242 on Build GRCh38
rs929645989 Chr.9: 27560840-27560840 on Build GRCh38
rs1006672180 Chr.9: 27560884-27560884 on Build GRCh38
rs1044297977 Chr.9: 27560807-27560807 on Build GRCh38
rs35844116 Chr.9: 27560953-27560954 on Build GRCh38
rs1032372883 Chr.9: 27561024-27561024 on Build GRCh38
rs549765761 Chr.9: 27561082-27561082 on Build GRCh38
rs111291984 Chr.9: 27561280-27561280 on Build GRCh38
rs999815895 Chr.9: 27561223-27561223 on Build GRCh38
rs138876353 Chr.9: 27560928-27560928 on Build GRCh38
rs902750563 Chr.9: 27561221-27561221 on Build GRCh38
rs981998006 Chr.9: 27561107-27561107 on Build GRCh38
rs542532552 Chr.9: 27561081-27561081 on Build GRCh38
rs1023432475 Chr.9: 27561076-27561076 on Build GRCh38
rs985445856 Chr.9: 27561265-27561265 on Build GRCh38
rs772869594 Chr.9: 27561054-27561062 on Build GRCh38
rs1005750788 Chr.9: 27561167-27561167 on Build GRCh38
rs905592444 Chr.9: 27561166-27561166 on Build GRCh38
rs761917781 Chr.9: 27561243-27561243 on Build GRCh38
rs946881734 Chr.9: 27560803-27560803 on Build GRCh38
rs972678222 Chr.9: 27560918-27560918 on Build GRCh38
rs555979063 Chr.9: 27561012-27561012 on Build GRCh38
rs984182373 Chr.9: 27561129-27561129 on Build GRCh38
rs1000210811 Chr.9: 27560811-27560811 on Build GRCh38
rs556170858 Chr.9: 27561011-27561011 on Build GRCh38
rs181689797 Chr.9: 27561092-27561092 on Build GRCh38
rs934054907 Chr.9: 27560792-27560792 on Build GRCh38
rs1017202533 Chr.9: 27560894-27560894 on Build GRCh38
rs868617075 Chr.9: 27560969-27560969 on Build GRCh38
rs918221652 Chr.9: 27560822-27560822 on Build GRCh38
rs1048122327 Chr.9: 27560851-27560851 on Build GRCh38
rs946820092 Chr.9: 27561044-27561044 on Build GRCh38
rs895522139 Chr.9: 27560826-27560826 on Build GRCh38
rs532281381 Chr.9: 27561264-27561264 on Build GRCh38
rs188774537 Chr.9: 27560795-27560795 on Build GRCh38
rs71746336 Chr.9: 27560802-27560806 on Build GRCh38
rs1051904740 Chr.9: 27561207-27561207 on Build GRCh38
rs922636790 Chr.9: 27560784-27560784 on Build GRCh38
rs73643416 Chr.9: 27560895-27560895 on Build GRCh38
rs1022632500 Chr.9: 27560873-27560873 on Build GRCh38
rs768575831 Chr.9: 27561132-27561132 on Build GRCh38
rs145979709 Chr.9: 27561140-27561140 on Build GRCh38
rs999914825 Chr.9: 27560989-27560989 on Build GRCh38
rs951369654 Chr.9: 27561288-27561288 on Build GRCh38
rs772319618 Chr.9: 27561300-27561300 on Build GRCh38
rs911282184 Chr.9: 27561010-27561010 on Build GRCh38
rs774359 Chr.9: 27561051-27561051 on Build GRCh38
rs557835944 Chr.9: 27561217-27561217 on Build GRCh38
rs896944970 Chr.9: 27561237-27561237 on Build GRCh38
rs970222939 Chr.9: 27560876-27560876 on Build GRCh38
rs12347222 Chr.9: 27560967-27560967 on Build GRCh38

View Full Product Details