Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCSTN
Entrez Gene ID
23385
Gene Name
nicastrin
Gene Aliases
ATAG1874
Chromosome Location
Chr.1: 160343056 - 160358952 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290184.1 NP_001277113.1 14 - 15 1813 81
NM_001290186.1 NP_001277115.1 10 - 11 1260 81
NM_015331.2 NP_056146.1 13 - 14 1674 81
XM_011509363.2 XP_011507665.1 13 - 14 1891 81
GenBank mRNA AF240468.1 - 13 - 14 1692 81
AK296153.1 - 8 - 9 959 81
AK299142.1 - 10 - 11 1147 81
AK314764.1 - 13 - 14 1581 81
AY359120.1 - 13 - 14 1551 81
BC047621.1 - 14 - 15 1770 81
D87442.1 - 13 - 14 1548 81

View Full Product Details