The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004559.4 | NP_004550.2 | 6 - 7 | 1071 | 83 |
NR_132737.1 | 5 - 6 | 889 | 83 | ||
GenBank mRNA | AK309996.1 | - | 6 - 7 | 708 | 83 |
BC000064.2 | - | 6 - 7 | 858 | 83 | |
BC002411.2 | - | 6 - 7 | 854 | 83 | |
BC010430.1 | - | 6 - 7 | 853 | 83 | |
BC013838.1 | - | 5 - 6 | 889 | 83 | |
BC015208.1 | - | 6 - 7 | 803 | 83 | |
BC018393.1 | - | 6 - 7 | 838 | 83 | |
BC020164.1 | - | 1 - 1 | 312 | 83 | |
BC038384.1 | - | 6 - 7 | 860 | 83 | |
BC065571.1 | - | 6 - 7 | 878 | 83 | |
BC070084.1 | - | 6 - 7 | 878 | 83 | |
BC071708.1 | - | 6 - 7 | 863 | 83 | |
BC072014.1 | - | 6 - 7 | 863 | 83 | |
BC090038.1 | - | 6 - 7 | 863 | 83 | |
BC098435.1 | - | 6 - 7 | 869 | 83 | |
BC106045.1 | - | 6 - 7 | 865 | 83 | |
J03827.1 | - | 6 - 7 | 866 | 83 | |
L28809.1 | - | 6 - 7 | 821 | 83 | |
M24070.1 | - | 6 - 7 | 860 | 83 |