Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32659422-32659965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DQB1
Entrez Gene ID
3119
Gene Name
major histocompatibility complex, class II, DQ beta 1
Gene Aliases
CELIAC1, HLA-DQB, HLA-DRB1, IDDM1
UniGene
Hs.409934
Gene Chromosome Location
Chr.6: 32659464-32666689 on Build GRCh38
Transcripts
NM_001243961.1 NM_001243962.1 NM_002123.4 NR_133907.1

Gene Symbol
HLA-DQB1-AS1
Entrez Gene ID
106480429
Gene Name
HLA-DQB1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.701274
Gene Chromosome Location
Chr.6: 32659880-32660729 on Build GRCh38
Transcripts
NM_001243961.1 NM_001243962.1 NM_002123.4 NR_133907.1

SNP Detail

SNP ID Chromosome Location
rs998941411 Chr.6: 32659460-32659460 on Build GRCh38
rs978411868 Chr.6: 32659861-32659861 on Build GRCh38
rs3204433 Chr.6: 32659891-32659891 on Build GRCh38
rs9273426 Chr.6: 32659657-32659657 on Build GRCh38
rs1130457 Chr.6: 32659590-32659590 on Build GRCh38
rs988508070 Chr.6: 32659894-32659894 on Build GRCh38
rs34153824 Chr.6: 32659850-32659850 on Build GRCh38
rs1049124 Chr.6: 32659936-32659936 on Build GRCh38
rs1130455 Chr.6: 32659628-32659628 on Build GRCh38
rs1064173 Chr.6: 32659703-32659703 on Build GRCh38
rs751600196 Chr.6: 32659612-32659612 on Build GRCh38
rs9273417 Chr.6: 32659575-32659575 on Build GRCh38
rs9273441 Chr.6: 32659796-32659796 on Build GRCh38
rs62998560 Chr.6: 32659758-32659758 on Build GRCh38
rs117789582 Chr.6: 32659826-32659826 on Build GRCh38
rs9273440 Chr.6: 32659784-32659784 on Build GRCh38
rs9273408 Chr.6: 32659444-32659444 on Build GRCh38
rs12918 Chr.6: 32659609-32659609 on Build GRCh38
rs9273444 Chr.6: 32659858-32659858 on Build GRCh38
rs9273430 Chr.6: 32659683-32659683 on Build GRCh38
rs34835883 Chr.6: 32659553-32659553 on Build GRCh38
rs9273432 Chr.6: 32659708-32659708 on Build GRCh38
rs9273419 Chr.6: 32659588-32659588 on Build GRCh38
rs1130456 Chr.6: 32659602-32659602 on Build GRCh38
rs9273418 Chr.6: 32659585-32659585 on Build GRCh38
rs63031662 Chr.6: 32659715-32659716 on Build GRCh38
rs9273425 Chr.6: 32659619-32659619 on Build GRCh38
rs9273424 Chr.6: 32659617-32659617 on Build GRCh38
rs756361095 Chr.6: 32659911-32659912 on Build GRCh38
rs1056316 Chr.6: 32659717-32659717 on Build GRCh38
rs9273413 Chr.6: 32659515-32659515 on Build GRCh38
rs914104471 Chr.6: 32659909-32659909 on Build GRCh38
rs994333262 Chr.6: 32659883-32659883 on Build GRCh38
rs112615708 Chr.6: 32659714-32659714 on Build GRCh38
rs191329273 Chr.6: 32659450-32659450 on Build GRCh38
rs765291761 Chr.6: 32659744-32659745 on Build GRCh38
rs9282075 Chr.6: 32659714-32659715 on Build GRCh38
rs1063355 Chr.6: 32659937-32659937 on Build GRCh38
rs1056315 Chr.6: 32659738-32659738 on Build GRCh38
rs9273439 Chr.6: 32659756-32659756 on Build GRCh38
rs9273414 Chr.6: 32659522-32659522 on Build GRCh38
rs1140365 Chr.6: 32659866-32659866 on Build GRCh38
rs376622401 Chr.6: 32659555-32659555 on Build GRCh38
rs6689 Chr.6: 32659923-32659923 on Build GRCh38
rs9273409 Chr.6: 32659458-32659458 on Build GRCh38
rs954251096 Chr.6: 32659504-32659504 on Build GRCh38
rs533116120 Chr.6: 32659641-32659641 on Build GRCh38
rs952726949 Chr.6: 32659781-32659781 on Build GRCh38
rs1130445 Chr.6: 32659843-32659843 on Build GRCh38
rs9273416 Chr.6: 32659544-32659544 on Build GRCh38
rs545181996 Chr.6: 32659806-32659806 on Build GRCh38
rs28724234 Chr.6: 32659758-32659758 on Build GRCh38
rs796901419 Chr.6: 32659617-32659619 on Build GRCh38
rs9273412 Chr.6: 32659486-32659486 on Build GRCh38
rs111951747 Chr.6: 32659465-32659465 on Build GRCh38
rs879233014 Chr.6: 32659753-32659753 on Build GRCh38
rs537937592 Chr.6: 32659560-32659560 on Build GRCh38
rs9273445 Chr.6: 32659869-32659869 on Build GRCh38
rs9273427 Chr.6: 32659664-32659664 on Build GRCh38
rs9273415 Chr.6: 32659533-32659533 on Build GRCh38
rs4993986 Chr.6: 32659875-32659875 on Build GRCh38
rs9273438 Chr.6: 32659746-32659746 on Build GRCh38
rs28724233 Chr.6: 32659669-32659669 on Build GRCh38
rs36205798 Chr.6: 32659836-32659836 on Build GRCh38
rs914304936 Chr.6: 32659842-32659842 on Build GRCh38
rs1056318 Chr.6: 32659673-32659673 on Build GRCh38
rs895632790 Chr.6: 32659572-32659572 on Build GRCh38
rs994229566 Chr.6: 32659701-32659701 on Build GRCh38
rs796465594 Chr.6: 32659588-32659590 on Build GRCh38
rs9273433 Chr.6: 32659712-32659712 on Build GRCh38
rs117067420 Chr.6: 32659877-32659877 on Build GRCh38
rs3204435 Chr.6: 32659856-32659856 on Build GRCh38
rs9273443 Chr.6: 32659823-32659823 on Build GRCh38
rs1130446 Chr.6: 32659840-32659840 on Build GRCh38
rs1020147809 Chr.6: 32659521-32659521 on Build GRCh38
rs1130451 Chr.6: 32659750-32659750 on Build GRCh38
rs9273410 Chr.6: 32659473-32659473 on Build GRCh38
rs9273437 Chr.6: 32659745-32659745 on Build GRCh38
rs9273429 Chr.6: 32659679-32659679 on Build GRCh38
rs9273442 Chr.6: 32659805-32659805 on Build GRCh38
rs9273423 Chr.6: 32659613-32659613 on Build GRCh38
rs9273411 Chr.6: 32659483-32659483 on Build GRCh38
rs9273436 Chr.6: 32659740-32659740 on Build GRCh38
rs3210187 Chr.6: 32659889-32659889 on Build GRCh38

View Full Product Details