The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308169.1 | NP_001295098.1 | 2 - 3 | 431 | 70 |
NM_001308172.1 | NP_001295101.1 | 3 - 4 | 616 | 70 | |
NM_001308954.1 | NP_001295883.1 | 4 - 5 | 998 | 70 | |
XM_017022923.1 | XP_016878412.1 | 4 - 5 | 591 | 70 | |
XM_017022924.1 | XP_016878413.1 | 3 - 4 | 501 | 70 | |
XM_017022925.1 | XP_016878414.1 | 2 - 3 | 388 | 70 | |
GenBank mRNA | AK055867.1 | - | 3 - 4 | 501 | 70 |
AK096039.1 | - | 4 - 5 | 857 | 70 | |
AK298359.1 | - | 2 - 3 | 320 | 70 | |
AK298434.1 | - | 3 - 4 | 470 | 70 | |
AK303774.1 | - | 2 - 3 | 316 | 70 | |
BC125176.1 | - | 3 - 4 | 453 | 70 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105069.1 | NP_001098539.1 | 3 - 4 | 562 | 70 |
NM_182617.3 | NP_872423.3 | 4 - 5 | 597 | 70 | |
XM_005255304.3 | XP_005255361.1 | 3 - 4 | 501 | 70 | |
XM_011545824.2 | XP_011544126.1 | 2 - 3 | 383 | 70 | |
XM_017023205.1 | XP_016878694.1 | 3 - 4 | 501 | 70 | |
GenBank mRNA | AB073604.1 | - | 4 - 5 | 528 | 70 |
AK129944.1 | - | 3 - 4 | 501 | 70 | |
AK289416.1 | - | 3 - 4 | 501 | 70 | |
AK309601.1 | - | 3 - 4 | 562 | 70 | |
DC362960.1 | - | 3 - 4 | 540 | 70 |