The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152864.3 | NP_690603.3 | 2 - 3 | 282 | 95 |
XM_005260192.1 | XP_005260249.1 | 2 - 3 | 244 | 95 | |
XM_005260194.3 | XP_005260251.1 | 2 - 3 | 703 | 95 | |
XM_011528527.1 | XP_011526829.1 | 2 - 3 | 244 | 95 | |
XM_011528528.2 | XP_011526830.1 | 2 - 3 | 706 | 95 | |
XM_011528529.2 | XP_011526831.1 | 2 - 3 | 692 | 95 | |
XM_017027636.1 | XP_016883125.1 | 2 - 3 | 214 | 95 | |
XM_017027637.1 | XP_016883126.1 | 2 - 3 | 696 | 95 | |
XM_017027638.1 | XP_016883127.1 | 2 - 3 | 702 | 95 | |
XM_017027639.1 | XP_016883128.1 | 2 - 3 | 688 | 95 | |
GenBank mRNA | AK126246.1 | - | 2 - 3 | 95 | |
AK225765.1 | - | 2 - 3 | 95 | ||
AK308958.1 | - | 2 - 3 | 95 | ||
BC041812.1 | - | 2 - 3 | 280 | 95 | |
BG708360.1 | - | 2 - 3 | 286 | 95 |