Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF12
Entrez Gene ID
7559
Gene Name
zinc finger protein 12
Gene Aliases
GIOT-3, HZF11, KOX3, ZNF325
Chromosome Location
Chr.7: 6688433 - 6706935 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006956.2 NP_008887.2 1 - 2 506 102
NM_016265.3 NP_057349.2 1 - 2 506 102
GenBank mRNA AB021643.1 - 1 - 2 506 102
AF505656.1 - 1 - 2 173 102
AK293925.1 - 2 - 3 152 102
BC051892.1 - 1 - 2 494 102
BC154414.1 - 1 - 2 494 102

View Full Product Details