The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173814.5 | NP_776175.2 | 1 - 2 | 280 | 98 |
XM_011521459.1 | XP_011519761.1 | 1 - 2 | 810 | 98 | |
XM_011521460.1 | XP_011519762.1 | 1 - 2 | 810 | 98 | |
XM_017022082.1 | XP_016877571.1 | 1 - 2 | 810 | 98 | |
GenBank mRNA | AY630258.1 | - | 1 - 2 | 140 | 98 |
CN352629.1 | - | 1 - 2 | 249 | 98 | |
CN352630.1 | - | 1 - 2 | 280 | 98 |