Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAIN1
Entrez Gene ID
642597
Gene Name
A kinase (PRKA) anchor inhibitor 1
Gene Aliases
C18orf42
Chromosome Location
Chr.18: 5143673 - 5199997 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011525728.2 XP_011524030.1 1 - 2 2550 129

View Full Product Details