Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXorf51B
Entrez Gene ID
100133053
Gene Name
chromosome X open reading frame 51B
Gene Aliases
-
Chromosome Location
Chr.X: 146809784 - 146810411 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244892.1 NP_001231821.1 1 - 2 147 146

Gene Symbol
CXorf51A
Entrez Gene ID
100129239
Gene Name
chromosome X open reading frame 51A
Gene Aliases
CXorf51
Chromosome Location
Chr.X: 146814104 - 146814731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144064.2 NP_001137536.1 1 - 2 147 146
GenBank mRNA AI954759.1 - 1 - 2 145

View Full Product Details