The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010977.2 | NP_001010977.1 | 3 - 4 | 426 | 72 |
XM_017020407.1 | XP_016875896.1 | 4 - 5 | 1320 | 72 | |
XM_017020408.1 | XP_016875897.1 | 4 - 5 | 1873 | 72 | |
GenBank mRNA | BC132748.1 | - | 3 - 4 | 426 | 72 |
BC132750.1 | - | 3 - 4 | 426 | 72 |